Methyl-4-bromopicolinate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64327-03
H64327-06
H64327-14
AAH64327-03EA
62.25
CAD
AAH64327-03
AAH64327-06
AAH64327-14
Methyl-4-bromopicolinate 97+%
Methyl 4-bromopicolinate
Methyl 4-bromopyridine-2-carboxylate, 97+%
Purity: 97+%
CAS No.: 29681-42-3
Molecular Formula: C7H6BrNO2
Formula Weight: 216.03
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 54-56°
Beilstein Reference: 473047
MDL No.: MFCD07374897
Formula:
C₇H₆BrNO₂ Melting Pt: 54…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD07374897 CAS Number: 29681-42-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...