5,6-Dichloropicolinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64320-03
H64320-06
AAH64320-03EA
184.86
CAD
AAH64320-03
AAH64320-06
AAH64320-MD
5,6-Dichloropicolinic acid 98%
5,6-Dichloropicolinic acid
5,6-Dichloropyridine-2-carboxylic acid, 98%
Purity: 98%
CAS No.: 88912-24-7
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Physical Form: Solid
Beilstein Reference: 4390497
MDL No.: MFCD13185546
Formula:
C₆H₃Cl₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13185546 CAS Number: 88912-24-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...