1-Bromo-4-fluorobutane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64317-03
H64317-06
H64317-14
AAH64317-03EA
65.73
CAD
AAH64317-03
AAH64317-06
AAH64317-14
1-Bromo-4-fluorobutane 97%
1-Bromo-4-fluorobutane
1-Bromo-4-fluorobutane, 97%
Purity: 97%
CAS No.: 462-72-6
Molecular Formula: C4H8BrF
Formula Weight: 155.01
Flash Point: 45°(113°F)
Physical Form: Liquid
Density: 1.444
Refractive Index: 1.4390
Appearance: Colorless
Boiling Point: 134-135°
Beilstein Reference: 1731414
UN No.: 1993
MDL No.: MFCD00209551
Formula:
BrH₂C(CH₂)₂CH₂F Boiling Pt: 134…135 °C Density: 1.444 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00209551 CAS Number: 462-72-6 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...