3-Bromo-4-methoxypyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64296-03
H64296-06
H64296-MD
AAH64296-03EA
181.7
CAD
AAH64296-03
AAH64296-06
AAH64296-MD
3-Bromo-4-methoxypyridine 97%
3-Bromo-4-methoxypyridine
3-Bromo-4-methoxypyridine, 97%
Purity: 97%
CAS No.: 82257-09-8
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Pale yellow
Beilstein Reference: 1423610
MDL No.: MFCD05664024
Formula:
C₆H₆BrNO MW: 188.02 Storage Temperature: Refrigerator |
MDL Number:
MFCD05664024 CAS Number: 82257-09-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...