(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(S)-(+)-Glycidyl nosylate, Glycidyl (S)-(+)-3-nitrobenzenesulfonate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulphonate, Glycidyl (S)-(+)-3-nitrobenzenesulphonate, (S)-Oxiran-2-ylmethyl 3-nitrobenzenesulfonate
H64289-06
H64289-14
AAH64289-03EA
43.54
CAD
AAH64289-03
AAH64289-06
AAH64289-14
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate 98%
(S)-Oxiran-2-ylmethyl-3-nitrobenzenesulfonate
Glycidyl (S)-(+)-3-nitrobenzenesulfonate, 98%
Purity: 98%
CAS No.: 115314-14-2
Molecular Formula: C9H9NO6S
Formula Weight: 259.23
Melting Point: 63-66°
Beilstein Reference: 4704457
MDL No.: MFCD00064582
Formula:
C9H9NO6S MW: 259.23 g/mol Melting Pt: 63…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064582 CAS Number: 115314-14-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...