Methyl-3-(4-hydroxyphenyl)propionate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe

H64255-14 H64255-22
AAH64255-14EA 141.41 CAD
AAH64255-14 AAH64255-22
Methyl-3-(4-hydroxyphenyl)propionate 98%
Methyl-3-(4-hydroxyphenyl)propionate

Methyl 3-(4-hydroxyphenyl)propionate, 98+%


Purity: 98+% CAS No.: 5597-50-2 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 113°(235°F) Physical Form: Powder Appearance: Off-white Melting Point: 39-41° lit. Boiling Point: 108°/11mm Beilstein Reference: 2211073 MDL No.: MFCD00071577

Formula: C10H12O3
Boiling Pt: 108 °C/11 mm
Melting Pt: 39…41 °C (lit.)
Flash Pt: >113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00071577
CAS Number: 5597-50-2

Order Now

Specification Test Results

Assay 98%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR