2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Bis(trifluoromethyl)-2-chloronitrobenzene
H64221-03
H64221-06
AAH64221-03EA
80.42
CAD
AAH64221-03
AAH64221-06
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene 98%
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene
2-Chloro-1-nitro-3,5-bis(trifluoromethyl)benzene, 98%
Purity: 98%
CAS No.: 654-55-7
Molecular Formula: C8H2ClF6NO2
Formula Weight: 293.55
Physical Form: Solid
Beilstein Reference: 2888862
MDL No.: MFCD04972677
Formula:
C₈H₂ClF₆NO₂ MW: 293.549 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04972677 CAS Number: 654-55-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...