1-Pyreneboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64211-03
H64211-06
AAH64211-03EA
76.49
CAD
AAH64211-03
AAH64211-06
1-Pyreneboronic acid 95%
1-Pyreneboronic acid
Pyrene-1-boronic acid, 95%
Purity: 95%
CAS No.: 164461-18-1
Molecular Formula: C16H11BO2
Formula Weight: 246.07
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 247-251° lit.
Beilstein Reference: 7134073
MDL No.: MFCD04974062
Formula:
C₁₆H₁₁BO₂ Melting Pt: 247…251 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD04974062 CAS Number: 164461-18-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...