3-Carboxyphenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64195-06
H64195-14
AAH64195-06EA
91.8
CAD
AAH64195-06
AAH64195-14
3-Carboxyphenylboronic acid pinacol ester 97%
3-Carboxyphenylboronic acid pinacol ester
3-Carboxybenzeneboronic acid pinacol ester, 97%
Purity: 97%
CAS No.: 269409-73-6
Molecular Formula: C13H17BO4
Formula Weight: 248.09
Physical Form: Powder
Appearance: White
Melting Point: 206-210° dec.
MDL No.: MFCD03411930
Formula:
C₁₃H₁₇BO₄ Melting Pt: 206…210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03411930 CAS Number: 269409-73-6 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...