Cyclohexylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclohexylboronic acid Pinacol ester, 2-Cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexane

H64178-03 H64178-06 H64178-14
AAH64178-03EA 59.25 CAD
AAH64178-03 AAH64178-06 AAH64178-14
Cyclohexylboronic acid pinacol ester 97%
Cyclohexylboronic acid pinacol ester

Cyclohexylboronic acid pinacol ester, 97%


Purity: 97% CAS No.: 87100-15-0 Molecular Formula: C12H23BO2 Formula Weight: 210.12 Physical Form: Liquid Appearance: Colorless MDL No.: MFCD04038749

Formula: C₁₂H₂₃BO₂
MW: 210.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038749
CAS Number: 87100-15-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR