3,5-Dibromo-4-methoxypyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64177-03
H64177-06
AAH64177-03EA
107.12
CAD
AAH64177-03
AAH64177-06
AAH64177-MD
3,5-Dibromo-4-methoxypyridine 98%
3,5-Dibromo-4-methoxypyridine
3,5-Dibromo-4-methoxypyridine, 98%
Purity: 98%
CAS No.: 650140-84-4
Molecular Formula: C6H5Br2NO
Formula Weight: 266.92
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD13185524
Formula:
C₆H₅Br₂NO₂ MW: 266.920 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185524 CAS Number: 25813-24-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...