5-(Trifluoromethyl)anthranilic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64141-03
H64141-MD
AAH64141-03EA
704.68
CAD
AAH64141-03
AAH64141-MD
5-(Trifluoromethyl)anthranilic acid 95%
5-(Trifluoromethyl)anthranilic acid
2-Amino-5-(trifluoromethyl)benzoic acid, 95%
Purity: 95%
CAS No.: 83265-53-6
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Flash Point:
Formula:
C₈H₆F₃NO₂ Melting Pt: 141…146 °C (dec.) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01569537 CAS Number: 83265-53-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...