3-Amino-1-adamantanol ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Aminoadamantan-1-ol

H64137-06 H64137-14
AAH64137-06EA 88.95 CAD
AAH64137-06 AAH64137-14 AAH64137-22
3-Amino-1-adamantanol ≥98%
3-Amino-1-adamantanol

Solid

Formula: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 265 °C (lit.)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01821204
CAS Number: 702-82-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR