4-Bromophthalimide ≥97%

Supplier: Thermo Scientific Chemicals
Warning

H64136-06 H64136-14
AAH64136-03EA 45.49 CAD
AAH64136-03 AAH64136-06 AAH64136-14
4-Bromophthalimide ≥97%
4-Bromophthalimide

4-Bromophthalimide, 97+%


Purity: 97+% CAS No.: 6941-75-9 Molecular Formula: C8H4BrNO2 Formula Weight: 226.03 Physical Form: Solid Appearance: Off-white Melting Point: 230-235° Beilstein Reference: 131235 MDL No.: MFCD00466049

Formula: C₈H₄BrNO₂
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00466049
CAS Number: 6941-75-9

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR