α,α,α,2,5-Pentafluorotoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,5-Difluorobenzotrifluoride

H64111-03 H64111-06
AAH64111-03EA 81.05 CAD
AAH64111-03 AAH64111-06
α,α,α,2,5-Pentafluorotoluene 98%
α,α,α,2,5-Pentafluorotoluene

2,5-Difluorobenzotrifluoride, 98%


Purity: 98% CAS No.: 393-38-4 Molecular Formula: C7H3F5 Formula Weight: 182.09 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.424 Refractive Index: 1.3940 Appearance: Colorless to pale yellow Melting Point: -14° Boiling Point: 107-108° Beilstein Reference: 2049494 MDL No.: MFCD00042494

Formula: C₇H₃F₅
Boiling Pt: 107…108 °C
Melting Pt: –14 °C
Density: 1.424
Storage Temperature: Ambient
MDL Number: MFCD00042494
CAS Number: 393-38-4

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR