2-(Trifluoromethyl)-5-pyridineboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid

H64106-03 H64106-06
AAH64106-03EA 167.48 CAD
AAH64106-03 AAH64106-06 AAH64106-MD
2-(Trifluoromethyl)-5-pyridineboronic acid 98%
2-(Trifluoromethyl)-5-pyridineboronic acid

2-(Trifluoromethyl)pyridine-5-boronic acid, 98%


Purity: 98% CAS No.: 868662-36-6 Molecular Formula: C6H5BF3NO2 Formula Weight: 190.92 Physical Form: Solid MDL No.: MFCD07375381

Formula: C₆H₅BF₃NO₂
MW: 190.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07375381
CAS Number: 868662-36-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR