3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64098-03
H64098-06
AAH64098-03EA
109.34
CAD
AAH64098-03
AAH64098-06
AAH64098-MD
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid 98%
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid
3-Cyclopropylmethoxy-4-difluoromethoxybenzoic acid, 98%
Purity: 98%
CAS No.: 162401-62-9
Molecular Formula: C12H12F2O4
Formula Weight: 258.22
Physical Form: Solid
Melting Point: 118-120°
Beilstein Reference: 11413145
MDL No.: MFCD04621687
Formula:
C₁₂H₁₂F₂O₄ Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD04621687 CAS Number: 162401-62-9 EINECS: 605-288-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...