2-(Trifluoromethyl)pyridine-5-carboxaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-(Trifluoromethyl)nicotinaldehyde
H64054-03
H64054-06
AAH64054-03EA
120.08
CAD
AAH64054-03
AAH64054-06
AAH64054-MD
2-(Trifluoromethyl)pyridine-5-carboxaldehyde 95%
2-(Trifluoromethyl)pyridine-5-carboxaldehyde
2-(Trifluoromethyl)pyridine-5-carboxaldehyde, 95%
Purity: 95%
CAS No.: 386704-12-7
Molecular Formula: C7H4F3NO
Formula Weight: 175.11
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Physical Form: Solid
Sensitivity: Air Sensitive
Melting Point: 52-56°
UN No.: 2811
MDL No.: MFCD01862647
Formula:
C7H4F3NO MW: 175.11 g/mol Melting Pt: 52…56 °C Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01862647 CAS Number: 386704-12-7 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...