2-Chloro-1-nitro-3-(trifluoromethyl)benzene 98%

Supplier: Thermo Scientific Chemicals
Warning

H64051-03 H64051-06
AAH64051-03EA 78.68 CAD
AAH64051-03 AAH64051-06 AAH64051-14
2-Chloro-1-nitro-3-(trifluoromethyl)benzene 98%
2-Chloro-1-nitro-3-(trifluoromethyl)benzene

2-Chloro-3-nitrobenzotrifluoride, 98%


Purity: 98% CAS No.: 39974-35-1 Molecular Formula: C7H3ClF3NO2 Formula Weight: 225.55 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 25-30° Beilstein Reference: 10429463 MDL No.: MFCD14525509

Formula: C₇H₃ClF₃NO₂
Melting Pt: 25…30 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD14525509
CAS Number: 39974-35-1
EINECS: 254-724-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR