5-Bromopyrimidine-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64041-06
H64041-MD
AAH64041-03EA
102.29
CAD
AAH64041-03
AAH64041-06
AAH64041-MD
5-Bromopyrimidine-2-carboxylic acid 97%
5-Bromopyrimidine-2-carboxylic acid
5-Bromopyrimidine-2-carboxylic acid, 98%
Purity: 98%
CAS No.: 37131-87-6
Molecular Formula: C5H3BrN2O2
Formula Weight: 203.00
Physical Form: Solid
Melting Point: 193-198°
Beilstein Reference: 121432
MDL No.: MFCD00496793
Formula:
C5H3BrN2O2 MW: 203.00 g/mol Melting Pt: 193…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00496793 CAS Number: 37131-87-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...