Boc-D-ABU-OH 95%
Supplier: Thermo Scientific Chemicals
H63991-03
H63991-06
H63991-14
AAH63991-03EA
62.25
CAD
AAH63991-03
AAH63991-06
AAH63991-14
Boc-D-ABU-OH 95%
(R)-2-(Boc-amino)butyric acid, 95%
Purity: 95%
CAS No.: 45121-22-0
Molecular Formula: C9H17NO4
Formula Weight: 203.24
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Yellow
MDL No.: MFCD00270335
Formula:
C₉H₁₇NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00270335 CAS Number: 45121-22-0 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...