4-(Methylthio)-1H-imidazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Methylthio)imidazole
H63990-03
H63990-MD
AAH63990-03EA
771.04
CAD
AAH63990-03
AAH63990-MD
4-(Methylthio)-1H-imidazole 97%
4-(Methylthio)-1H-imidazole
4-(Methylthio)imidazole, 97%
Purity: 97%
CAS No.: 83553-60-0
Molecular Formula: C4H6N2S
Formula Weight: 114.17
Storage Temperature: 2°C to 8°C
MDL No.: MFCD03844702
Formula:
C₄H₆N₂S MW: 114.166 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03844702 CAS Number: 83553-60-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...