L-Aspartic aciddiethyl ester hydrochloride 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Asp(OEt)-OEt·HCl, H-Asp(OEt)-OEt.HCl, Diethyl L-Aspartate hydrochloride, L-Aspartic acid diethyl ester hydrochloride
H63974-06
H63974-14
H63974-22
AAH63974-06EA
51.51
CAD
AAH63974-06
AAH63974-14
AAH63974-22
L-Aspartic aciddiethyl ester hydrochloride 98%
L-Aspartic aciddiethyl ester hydrochloride
L-Aspartic acid diethyl ester hydrochloride, 98%
Purity: 98%
CAS No.: 16115-68-7
Molecular Formula: C8H15NO4.HCl
Formula Weight: 225.67
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00038910
Formula:
C₈H₁₆ClNO₄ MW: 225.669 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038910 CAS Number: 16115-68-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...