Z-Glu-OtBu 95%
Supplier: Thermo Scientific Chemicals
H63954-03
H63954-06
AAH63954-03EA
117.08
CAD
AAH63954-03
AAH63954-06
AAH63954-MD
Z-Glu-OtBu 95%
Z-Glu-OtBu
N-Benzyloxycarbonyl-L-glutamic acid 1-tert-butyl ester, 95%
Purity: 95%
CAS No.: 5891-45-2
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Storage Temperature: 2°C to 8°C
Physical Form: Solid
MDL No.: MFCD00077016
Formula:
C₁₇H₂₃NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD00077016 CAS Number: 5891-45-2 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...