2-bromo-5-methylthiazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-5-methylthiazole

H63951-03 H63951-06
AAH63951-03EA 56.72 CAD
AAH63951-03 AAH63951-06
2-bromo-5-methylthiazole 98%
2-bromo-5-methylthiazole

2-Bromo-5-methylthiazole, 98%


Purity: 98% CAS No.: 41731-23-1 Molecular Formula: C4H4BrNS Formula Weight: 178.05 MDL No.: MFCD08460610

Formula: C₄H₄BrNS
MW: 178.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460610
CAS Number: 41731-23-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR