2,3-Dimethylphenylacetic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63947-03
H63947-06
AAH63947-03EA
92.27
CAD
AAH63947-03
AAH63947-06
AAH63947-MD
2,3-Dimethylphenylacetic acid 95%
2,3-Dimethylphenylacetic acid
2,3-Dimethylphenylacetic acid, 95%
Purity: 95%
CAS No.: 30981-98-7
Molecular Formula: C10H12O2
Formula Weight: 164.2
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD03844736
Formula:
C₁₀H₁₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03844736 CAS Number: 30981-98-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...