3-(3-Pyridyl)-L-alanine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-2-Amino-3-(pyridin-3-yl)propanoic acid, H-β-(3-Pyridyl)-Ala-OH, H-3-Pal-OH

H63941-03 H63941-06 H63941-MD
AAH63941-03EA 265.44 CAD
AAH63941-03 AAH63941-06 AAH63941-MD
3-(3-Pyridyl)-L-alanine 98%
3-(3-Pyridyl)-L-alanine

3-(3-Pyridyl)-L-alanine, 98%


Purity: 98% CAS No.: 64090-98-8 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Physical Form: Powder Appearance: Off-white Melting Point: 248-252° dec. Beilstein Reference: 4351583 MDL No.: MFCD00079676

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 248…252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00079676
CAS Number: 64090-98-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR