(±)-β-Aminophenethyl alcohol 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

H63932-06 H63932-14
AAH63932-06EA 53.4 CAD
AAH63932-06 AAH63932-14 AAH63932-22
(±)-β-Aminophenethyl alcohol 95%
(±)-β-Aminophenethyl alcohol

DL-Phenylglycinol, 95%


Purity: 95% CAS No.: 7568-92-5 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Solid Appearance: Off-white Melting Point: 49-53° Beilstein Reference: 971219 MDL No.: MFCD00130145

Formula: C₈H₁₁NO
Melting Pt: 49…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00130145
CAS Number: 7568-92-5
UN: 3263
ADR: "8",III

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR