Fmoc-Inp-OH 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-piperidinecarboxylic acid, 1-Fmoc-isonipecotic acid, 1-[(9H-Fluoren-9-ylmethoxy)carbonyl]isonipecotic acid, Fmoc-isonipecotic acid, 1-Fmoc-4-piperidinecarboxylic acid, Fmoc-Isn-OH, 1-Fmoc-piperidine-4-carboxylic acid

H63915-06 H63915-14
AAH63915-03EA 39.22 CAD
AAH63915-03 AAH63915-06 AAH63915-14
Fmoc-Inp-OH 98%
Fmoc-Inp-OH

1-Fmoc-piperidine-4-carboxylic acid, 98%


Purity: 98% CAS No.: 148928-15-8 Molecular Formula: C21H21NO4 Formula Weight: 351.4 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: ca 190° dec Beilstein Reference: 7722550 MDL No.: MFCD00273475

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Melting Pt: ca 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00273475
CAS Number: 148928-15-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR