N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine
H63910-03
H63910-06
AAH63910-03EA
94.48
CAD
AAH63910-03
AAH63910-06
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
N-Fmoc-3-cyclohexyl-L-alanine, 98%
Purity: 98%
CAS No.: 135673-97-1
Molecular Formula: C24H27NO4
Formula Weight: 393.48
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 7052264
MDL No.: MFCD00065614
Formula:
C₂₄H₂₇NO₄ MW: 393.48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00065614 CAS Number: 135673-97-1 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...