H-Tyr(BZL)-OMe·HCl 98%

Supplier: Thermo Scientific Chemicals

Synonyms: O-Benzyl-L-tyrosine methyl ester hydrochloride, H-Tyr(Bzl)-OMe·HCl

H63908-03 H63908-06
AAH63908-03EA 51.98 CAD
AAH63908-03 AAH63908-06
H-Tyr(BZL)-OMe·HCl 98%
H-Tyr(BZL)-OMe·HCl

O-Benzyl-L-tyrosine methyl ester hydrochloride, 98%


Purity: 98% CAS No.: 34805-17-9 Molecular Formula: C17H19NO3.HCl Formula Weight: 321.8 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD00080888

Formula: C₁₇H₂₀ClNO₃
MW: 321.801 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080888
CAS Number: 34805-17-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR