2-Bromo-4,5-difluorophenylacetic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(2-Bromo-4,5-difluorophenyl)acetic acid
H63902-03
H63902-06
AAH63902-03EA
143.15
CAD
AAH63902-03
AAH63902-06
AAH63902-MD
2-Bromo-4,5-difluorophenylacetic acid 98%
2-Bromo-4,5-difluorophenylacetic acid
2-Bromo-4,5-difluorophenylacetic acid, 98%
Purity: 98%
CAS No.: 883502-07-6
Molecular Formula: C8H5BrF2O2
Formula Weight: 251.03
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD04973423
Formula:
C₈H₅BrF₂O₂ MW: 251.027 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973423 CAS Number: 883502-07-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...