Fmoc-N-Me-D-Leu-OH 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Fmoc-N-methyl-D-leucine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methylpentanoic acid, Fmoc-D-N-Me-Leu-OH
H63895-03
H63895-06
H63895-14
AAH63895-03EA
159.58
CAD
AAH63895-03
AAH63895-06
AAH63895-14
Fmoc-N-Me-D-Leu-OH 97%
Fmoc-N-Me-D-Leu-OH
N-Fmoc-N-methyl-D-leucine, 97%
Purity: 97%
CAS No.: 103478-63-3
Molecular Formula: C22H25NO4
Formula Weight: 367.44
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 7350417
MDL No.: MFCD00235877
Formula:
C₂₂H₂₅NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00235877 CAS Number: 103478-63-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...