4-(4-Fluorophenyl)piperidin-4-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

H63892-03 H63892-06
AAH63892-03EA 133.98 CAD
AAH63892-03 AAH63892-06 AAH63892-MD
4-(4-Fluorophenyl)piperidin-4-ol 97%
4-(4-Fluorophenyl)piperidin-4-ol

4-(4-Fluorophenyl)-4-hydroxypiperidine, 97%


Purity: 97% CAS No.: 3888-65-1 Molecular Formula: C11H14FNO Formula Weight: 195.23 Physical Form: Solid MDL No.: MFCD00023146

Formula: C₁₁H₁₄FNO
Storage Temperature: Ambient
MDL Number: MFCD00023146
CAS Number: 3888-65-1
EINECS: 223-431-8

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR