3-Chloro-6-iodopyridazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63879-03
H63879-06
AAH63879-03EA
95.27
CAD
AAH63879-03
AAH63879-06
3-Chloro-6-iodopyridazine 97%
3-Chloro-6-iodopyridazine
3-Chloro-6-iodopyridazine, 97%
Purity: 97%
CAS No.: 135034-10-5
Molecular Formula: C4H2ClIN2
Formula Weight: 240.43
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 110-120°
MDL No.: MFCD08275187
Formula:
C₄H₂ClIN₂ Melting Pt: 110…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD08275187 CAS Number: 135034-10-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...