Ac-Glu(OtBu)-OH 95%
Supplier: Thermo Scientific Chemicals
H63871-03
H63871-06
AAH63871-03EA
100.49
CAD
AAH63871-03
AAH63871-06
AAH63871-MD
Ac-Glu(OtBu)-OH 95%
Ac-Glu(OtBu)-OH
N-Acetyl-L-glutamic acid 5-tert-butyl ester, 95%
Purity: 95%
CAS No.: 84192-88-1
Molecular Formula: C11H19NO
Formula Weight: 245.27
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00236754
Formula:
C₁₁H₁₉NO₅ Storage Temperature: Ambient |
MDL Number:
MFCD00236754 CAS Number: 84192-88-1 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...