1-(Methylsulfonyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H63870-06 H63870-14 H63870-22
AAH63870-06EA 62.25 CAD
AAH63870-06 AAH63870-14 AAH63870-22
1-(Methylsulfonyl)piperazine 97%
1-(Methylsulphonyl)piperazine

1-(Methylsulfonyl)piperazine, 97%


Purity: 97% CAS No.: 55276-43-2 Molecular Formula: C5H12N2O2S Formula Weight: 164.23 Physical Form: Solid MDL No.: MFCD02153225

Formula: C₅H₁₂N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD02153225
CAS Number: 55276-43-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR