H-Gln-OtBu·HCl 98%

Supplier: Thermo Scientific Chemicals

Synonyms: L-Glutamine tert-butyl ester hydrochloride, H-Gln-OtBu.HCl, H-Gln-OtBu·HCl

H63868-03 H63868-06
AAH63868-03EA 173.8 CAD
AAH63868-03 AAH63868-06 AAH63868-MD
H-Gln-OtBu·HCl 98%
H-Gln-OtBu·HCl

L-Glutamine tert-butyl ester hydrochloride, 98%


Purity: 98% CAS No.: 39741-62-3 Molecular Formula: C9H18N2O3.HCl Formula Weight: 238.71 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White MDL No.: MFCD00039081

Formula: C₉H₁₉ClN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00039081
CAS Number: 39741-62-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR