2-Bromo-5-formylpyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63867-03
H63867-06
H63867-14
AAH63867-03EA
47.4
CAD
AAH63867-03
AAH63867-06
AAH63867-14
2-Bromo-5-formylpyridine 95%
2-Bromo-5-formylpyridine
2-Bromopyridine-5-carboxaldehyde, 95%
Purity: 95%
CAS No.: 149806-06-4
Molecular Formula: C6H4BrNO
Formula Weight: 186.01
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Pale yellow
Melting Point: 104-110° dec
MDL No.: MFCD04115419
Formula:
C₆H₄BrNO Melting Pt: 104…110 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04115419 CAS Number: 149806-06-4 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...