BippyPhos 97%
Supplier: Thermo Scientific Chemicals
H63864-03
H63864-MD
AAH63864-03EA
417.12
CAD
AAH63864-03
AAH63864-MD
BippyPhos 97%
BippyPhos
5-Di-tert-butylphosphino-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 97%
Purity: 97%
CAS No.: 894086-00-1
Molecular Formula: C32H35N4P
Formula Weight: 506.62
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 169-173°
MDL No.: MFCD09038440
Formula:
C₃₂H₃₅N₄P Melting Pt: 169…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD09038440 CAS Number: 894086-00-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...