4-(4-Fluorobenzyl)piperidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63860-03
H63860-06
AAH63860-03EA
70.94
CAD
AAH63860-03
AAH63860-06
AAH63860-14
4-(4-Fluorobenzyl)piperidine hydrochloride 97%
4-(4-Fluorobenzyl)piperidine hydrochloride
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Formula:
C₁₂H₁₆FN·ClH Storage Temperature: Ambient |
MDL Number:
MFCD03840140 CAS Number: 92822-03-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...