N-(ɑ)-Boc-L-lysine 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-6-Amino-2-((tert-butoxycarbonyl)amino)hexanoic acid, Nalpha-Boc-L-lysine, Boc-Lys-OH
H63859-06
H63859-14
H63859-22
AAH63859-06EA
165.9
CAD
AAH63859-06
AAH63859-14
AAH63859-22
N-(ɑ)-Boc-L-lysine 97%
N-(alpha)-Boc-L-lysine
N^a-Boc-L-lysine, 97%
Purity: 97%
CAS No.: 13734-28-6
Molecular Formula: C11H22N2O4
Formula Weight: 246.30
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: ca 205° dec
Beilstein Reference: 4252546
Optical Rotation: +22°(c=2 in methanol)
MDL No.: MFCD00038203
Formula:
C11H22N2O4 MW: 246.3 g/mol Melting Pt: ca 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038203 CAS Number: 13734-28-6 EINECS: 237-303-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...