4-(2-Fluorobenzyl)piperidine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63849-03
H63849-06
AAH63849-03EA
385.52
CAD
AAH63849-03
AAH63849-06
AAH63849-MD
4-(2-Fluorobenzyl)piperidine hydrochloride 97%
4-(2-Fluorobenzyl)piperidine hydrochloride
4-(2-Fluorobenzyl)piperidine hydrochloride, 97%
Purity: 97%
CAS No.: 193357-21-0
Molecular Formula: C12H16FN.HCl
Formula Weight: 229.72
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
MDL No.: MFCD09787475
Formula:
C₁₂H₁₇ClFN Storage Temperature: Ambient |
MDL Number:
MFCD09787475 CAS Number: 193357-21-0 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...