2-Iodo-5-trifluoromethylpyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

H63848-03 H63848-06
AAH63848-03EA 155.79 CAD
AAH63848-03 AAH63848-06 AAH63848-MD
2-Iodo-5-trifluoromethylpyridine 96%
2-Iodo-5-trifluoromethylpyridine

2-Iodo-5-(trifluoromethyl)pyridine, 96%


Purity: 96% CAS No.: 100366-75-4 Molecular Formula: C6H3F3IN Formula Weight: 272.99 MDL No.: MFCD08741351

Formula: C₆H₃F₃IN
Storage Temperature: Ambient
MDL Number: MFCD08741351
CAS Number: 100366-75-4

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR