1-[3-(Dimethylamino)propyl]-3-ethylurea 97%

Supplier: Thermo Scientific Chemicals
Warning

H63845-03 H63845-06 H63845-MD
AAH63845-03EA 263.86 CAD
AAH63845-03 AAH63845-06 AAH63845-MD
1-[3-(Dimethylamino)propyl]-3-ethylurea 97%
1-[3-(Dimethylamino)propyl]-3-ethylurea

1-[3-(Dimethylamino)propyl]-3-ethylurea, 97%


Purity: 97% CAS No.: 32897-26-0 Molecular Formula: C8H19N3O Formula Weight: 173.26 Beilstein Reference: 4961600 MDL No.: MFCD00027752

Formula: C₈H₁₉N₃O
MW: 173.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027752
CAS Number: 32897-26-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR