2-Thiopheneboronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
H63819-03
H63819-06
H63819-14
AAH63819-03EA
52.46
CAD
AAH63819-03
AAH63819-06
AAH63819-14
2-Thiopheneboronic acid pinacol ester 98%
2-Thiopheneboronic acid pinacol ester
Thiophene-2-boronic acid pinacol ester, 98%
Purity: 98%
CAS No.: 193978-23-3
Molecular Formula: C10H17BO3S
Formula Weight: 228.12
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 65-70°
MDL No.: MFCD05663878
Formula:
C₁₀H₁₅BO₂S Melting Pt: 65…70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD05663878 CAS Number: 193978-23-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...