H-Aib-OtBu·HCl 98%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl 2-aminoisobutyrate hydrochloride, tert-Butyl 2-amino-2-methylpropanoate hydrochloride

H63817-03 H63817-06
AAH63817-03EA 224.36 CAD
AAH63817-03 AAH63817-06 AAH63817-MD
H-Aib-OtBu·HCl 98%
H-Aib-OtBu·HCl

tert-Butyl 2-aminoisobutyrate hydrochloride, 98%


Purity: 98% CAS No.: 4512-32-7 Molecular Formula: C8H17NO2.HCl Formula Weight: 195.69 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD08272284

Formula: C₈H₁₇NO₂
MW: 195.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272284
CAS Number: 84758-81-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR