(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene 97%
Supplier: Thermo Scientific Chemicals
H63816-MC
H63816-ME
AAH63816-MCEA
128.96
CAD
AAH63816-MC
AAH63816-ME
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene 97%
(S)-1-(Diphenylphosphino)-2-[(S)-4-isopropyloxazolin-2-yl]ferrocene
(S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine, 97%
Purity: 97%
CAS No.: 163169-29-7
Molecular Formula: C28H28FeNOP
Formula Weight: 481.35
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Orange
Melting Point: 158-162°
Optical Rotation: 366°(c=0.1 in chloroform)
MDL No.: MFCD15144869
Formula:
C₂₈H₂₈FeNOP Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD15144869 CAS Number: 163169-29-7 UN: 3335 ADR: "9",III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...