O-tert-Butyl-L-serine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Ser(tBu)-OH
H63812-03
H63812-06
AAH63812-03EA
72.68
CAD
AAH63812-03
AAH63812-06
O-tert-Butyl-L-serine 98%
O-tert-Butyl-L-serine
O-tert-Butyl-D-serine, 98%
Purity: 98%
CAS No.: 18822-58-7
Molecular Formula: C7H15NO3
Formula Weight: 161.2
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White
Melting Point: 220° dec
Beilstein Reference: 2500118
Optical Rotation: -16°(c=1 in water)
MDL No.: MFCD00066088
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol Melting Pt: 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00066088 CAS Number: 18822-58-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...