3-Boc-aminoethylazetidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63797-03
H63797-MD
AAH63797-03EA
1039.64
CAD
AAH63797-03
AAH63797-MD
3-Boc-aminoethylazetidine 95%
3-Boc-aminoethylazetidine
3-[2-(Boc-amino)ethyl]azetidine, 95%
Purity: 95%
CAS No.: 162696-31-3
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 84-86°
MDL No.: MFCD09878605
Formula:
C₁₀H₂₀N₂O₂ Melting Pt: 84…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD09878605 CAS Number: 162696-31-3 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...